CH6371381323 Chem Impex International, Inc.

(S)-3-Amino-3-(4-fluorophenyl)propionic acid

  • MDL number MFCD03840458
  • Cas Number : 151911-33-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym L-beta-Phe(4-F)-OH
Purity 98
Storage temp. Store at 0-8
Physical Form Off-white to pale yellow powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-s-i-3-amino-3-4-fluorophenyl-propionic-acid/6595?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-s-i-3-amino-3-4-fluorophenyl-propionic-acid/6595?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart