CH6371381327 Chem Impex International, Inc.

(-)-(1S,4R)-4-Aminocyclopent-2-enecarboxylic acid

  • MDL number MFCD00211286
  • Cas Number : 168471-40-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (-)-(1S,4R)-gamma-Homocycloleu-2-ene
Purity 99
Storage temp. Store at 0-8
Physical Form Off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/1-i-s-i-4-i-r-i-4-aminocyclopent-2-enecarboxylic-acid/6599?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/1-i-s-i-4-i-r-i-4-aminocyclopent-2-enecarboxylic-acid/6599?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart