CH6371381330 Chem Impex International, Inc.

4-Methyl-D-beta-homophenylalanine hydrochloride

  • MDL number MFCD01860927
  • Cas Number : 177839-85-9
  • Molecular Weight : 229.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 252-258 °C
Purity 98
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/4-methyl-d-beta-homophenylalanine-hydrochloride/6602?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/4-methyl-d-beta-homophenylalanine-hydrochloride/6602?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart