CH6371381331 Chem Impex International, Inc.

(R)-2-Tetrahydroisoquinoline acetic acid hydrochloride

  • MDL number MFCD01860932
  • Cas Number : 187218-03-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 206-212 °C
Purity 98
Storage temp. Store at 0-8 °C
Physical Form Off-white powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-r-i-2-tetrahydroisoquinoline-acetic-acid-hydrochloride/6603?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-r-i-2-tetrahydroisoquinoline-acetic-acid-hydrochloride/6603?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart