CH6371381332 Chem Impex International, Inc.

Boc-(2-thienyl)-L-b-homoalanine

  • MDL number MFCD01861081
  • Cas Number : 190190-47-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Boc-(2-thienyl)-L-beta-homoalanine
Purity 0.99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-2-thienyl-l-beta-homoalanine/6604?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-2-thienyl-l-beta-homoalanine/6604?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart