CH6371381333 Chem Impex International, Inc.

Boc-(3-benzothienyl)-D-beta-homoalanine

  • MDL number MFCD01860943
  • Cas Number : 190190-48-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 123 - 127 °C
Purity 99
Storage temp. Store at 0-8°C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-3-benzothienyl-d-ß-homoalanine/6605?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-3-benzothienyl-d-ß-homoalanine/6605?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart