CH6371381334 Chem Impex International, Inc.

Boc-(1-naphthyl)-D-beta-homoalanine

  • MDL number MFCD01076281
  • Cas Number : 190190-49-9
  • Molecular Weight : 329.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8°C
Physical Form White solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-1-naphthyl-d-ß-homoalanine/6606?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-1-naphthyl-d-ß-homoalanine/6606?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart