CH6371381349 Chem Impex International, Inc.

(+)-(1S,3R)-N-Fmoc-3-aminocyclopentane carboxylic acid

  • MDL number MFCD01311757
  • Cas Number : 220497-66-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (+)-(1S,3R)-N-Fmoc-beta-homocycloleucine
Purity 0.95
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/1-i-s-i-3-i-r-i-i-n-i-fmoc-3-aminocyclopentane-carboxylic-acid/6630?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/1-i-s-i-3-i-r-i-i-n-i-fmoc-3-aminocyclopentane-carboxylic-acid/6630?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart