CH6371381350 Chem Impex International, Inc.

(-)-(1R,3S)-N-Fmoc-3-aminocyclopentane carboxylic acid

  • MDL number MFCD01311758
  • Cas Number : 220497-67-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (-)-(1R,3S)-N-Fmoc-beta-homocycloleucine
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/1-i-r-i-3-i-s-i-i-n-i-fmoc-3-aminocyclopentane-carboxylic-acid/6631?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/1-i-r-i-3-i-s-i-i-n-i-fmoc-3-aminocyclopentane-carboxylic-acid/6631?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart