CH6371381352 Chem Impex International, Inc.

Fmoc-D-alpha-(5-bromothienyl)alanine

  • MDL number MFCD01311772
  • Cas Number : 220497-83-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8°C
Physical Form White solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-d-a-5-bromothienyl-alanine/6633?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-d-a-5-bromothienyl-alanine/6633?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart