CH6371381358 Chem Impex International, Inc.

(R)-3-Amino-3-(3-chlorophenyl)propionic acid

  • MDL number MFCD04113654
  • Cas Number : 262429-49-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8°C
Physical Form White to off-white solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-r-i-3-amino-3-3-chlorophenyl-propionic-acid/6640?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-r-i-3-amino-3-3-chlorophenyl-propionic-acid/6640?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart