CH6371381360 Chem Impex International, Inc.

2-Chloro-D-b-homophenylalanine hydrochloride

  • MDL number MFCD01860898
  • Cas Number : 268734-28-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym D-beta-HomoPhe(2-Cl)-OH HCl
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/2-chloro-d-beta-homophenylalanine-hydrochloride/6642?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/2-chloro-d-beta-homophenylalanine-hydrochloride/6642?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart