CH6371381362 Chem Impex International, Inc.

3,4-Dichloro-D-beta-homophenylalanine hydrochloride

  • MDL number MFCD01860948
  • Cas Number : 269396-55-6
  • Molecular Weight : 284.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 219-226 °C
Purity 98
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/3-4-dichloro-d-beta-homophenylalanine-hydrochloride/6648?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/3-4-dichloro-d-beta-homophenylalanine-hydrochloride/6648?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart