CH6371381366 Chem Impex International, Inc.

(4-Pyridyl)-D-beta-homoalanine dihydrochloride

  • MDL number MFCD01860960
  • Cas Number : 269396-67-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 267-271 °C (dec.)
Purity 98
Storage temp. Store at 0-8 °C
Physical Form Off-white powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/4-pyridyl-d-beta-homoalanine-dihydrochloride/6657?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/4-pyridyl-d-beta-homoalanine-dihydrochloride/6657?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart