CH6371381376 Chem Impex International, Inc.

Fmoc-(R)-3-amino-5-phenylpentanoic acid

  • MDL number MFCD01860931
  • Cas Number : 269398-87-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Fmoc-D-beta-Nva(5-phenyl)-OH
Purity 98
Storage temp. Store at 0-8
Physical Form White solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-i-r-i-3-amino-5-phenylpentanoic-acid/6674?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-i-r-i-3-amino-5-phenylpentanoic-acid/6674?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart