CH6371381383 Chem Impex International, Inc.

(3-Benzothienyl)-D-b-homoalanine hydrochloride

  • MDL number MFCD01860942
  • Cas Number : 269398-95-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 221-227 �C
Synonym D-beta-HomoAla(3-benzothienyl)-OH HCl
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/3-benzothienyl-d-beta-homoalanine-hydrochloride/6682?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/3-benzothienyl-d-beta-homoalanine-hydrochloride/6682?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart