CH6371381384 Chem Impex International, Inc.

3-Trifluoromethyl-D-beta-homophenylalanine hydrochloride

  • MDL number MFCD01860968
  • Cas Number : 269726-73-0
  • Molecular Weight : 283.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 236-242 °C
Purity 98
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/3-trifluoromethyl-d-beta-homophenylalanine-hydrochloride/6684?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/3-trifluoromethyl-d-beta-homophenylalanine-hydrochloride/6684?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart