CH6371381391 Chem Impex International, Inc.

(2-Thienyl)-D-b-homoalanine hydrochloride

  • MDL number MFCD01860983
  • Cas Number : 269726-88-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 166-172 �C
Synonym D-beta-HomoAla(2-thienyl)-OH HCl
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/2-thienyl-d-beta-homoalanine-hydrochloride/6699?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/2-thienyl-d-beta-homoalanine-hydrochloride/6699?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart