CH6371381395 Chem Impex International, Inc.

(3-Thienyl)-D-beta-homoalanine hydrochloride

  • MDL number MFCD01860986
  • Cas Number : 269726-91-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 196-202 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/3-thienyl-d-beta-homoalanine-hydrochloride/6704?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/3-thienyl-d-beta-homoalanine-hydrochloride/6704?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart