CH6371381405 Chem Impex International, Inc.

3-Methyl-L-beta-homophenylalanine hydrochloride

  • MDL number MFCD01861022
  • Cas Number : 270062-92-5
  • Molecular Weight : 229.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature RT
Country of Origin US
Product Link http://www.chemimpex.com/product/productinfo/3-methyl-l-beta-homophenylalanine-hydrochloride/6717?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/3-methyl-l-beta-homophenylalanine-hydrochloride/6717?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart