CH6371381408 Chem Impex International, Inc.

Fmoc-(S)-2-tetrahydroisoquinoline acetic acid

  • MDL number MFCD01861032
  • Cas Number : 270062-99-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Fmoc-Tqa-OH
Purity 0.99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-i-s-i-2-tetrahydroisoquinoline-acetic-acid/6724?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-i-s-i-2-tetrahydroisoquinoline-acetic-acid/6724?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart