CH6371381412 Chem Impex International, Inc.

L-b-HomoAla(3-benzothienyl)-OH�

  • MDL number MFCD01861040
  • Cas Number : 270063-44-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 222-228 �C
Synonym (3-Benzothienyl)-L-beta-homoalanine hydrochloride
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/l-beta-homoala-3-benzothienyl-oh-font-size-2-face-symbol-middot-font-hcl/6732?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/l-beta-homoala-3-benzothienyl-oh-font-size-2-face-symbol-middot-font-hcl/6732?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart