CH6371381414 Chem Impex International, Inc.

2,4-Dichloro-L-b-homophenylalanine hydrochloride

  • MDL number MFCD01861043
  • Cas Number : 270063-47-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 195-199 �C (dec.)
Synonym L-beta-HomoPhe(2,4-DiCl)-OH�
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/2-4-dichloro-l-beta-homophenylalanine-hydrochloride/6735?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/2-4-dichloro-l-beta-homophenylalanine-hydrochloride/6735?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart