CH6371381420 Chem Impex International, Inc.

Fmoc-(4-pyridyl)-L-beta-homoalanine

  • MDL number MFCD01861059
  • Cas Number : 270065-69-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 172-178 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-4-pyridyl-l-beta-homoalanine/6747?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-4-pyridyl-l-beta-homoalanine/6747?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart