CH6371381424 Chem Impex International, Inc.

2-Trifluoromethyl-L-b-homophenylalanine hydrochloride

  • MDL number MFCD01861063
  • Cas Number : 270065-73-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 200-206 �C
Synonym L-beta-HomoPhe(2-CF3)-OH HCl
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/2-trifluoromethyl-l-beta-homophenylalanine-hydrochloride/6751?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/2-trifluoromethyl-l-beta-homophenylalanine-hydrochloride/6751?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart