CH6371381426 Chem Impex International, Inc.

3-Trifluoromethyl-L-beta-homophenylalanine hydrochloride

  • MDL number MFCD01861066
  • Cas Number : 270065-76-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 238-244 °C
Purity 98
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/3-trifluoromethyl-l-beta-homophenylalanine-hydrochloride/6754?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/3-trifluoromethyl-l-beta-homophenylalanine-hydrochloride/6754?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart