CH6371381427 Chem Impex International, Inc.

Fmoc-3-trifluoromethyl-L-beta-homophenylalanine

  • MDL number MFCD01861068
  • Cas Number : 270065-78-6
  • Molecular Weight : 469.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 149-155 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-3-trifluoromethyl-l-beta-homophenylalanine/6756?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-3-trifluoromethyl-l-beta-homophenylalanine/6756?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart