CH6371381429 Chem Impex International, Inc.

2-Cyano-L-beta-homophenylalanine hydrochloride

  • MDL number MFCD01861072
  • Cas Number : 270065-82-2
  • Molecular Weight : 240.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 202-208 °C
Purity 98
Storage temp. Store at 0-8°C
Physical Form White Powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/2-cyano-l-beta-homophenylalanine-hydrochloride/6760?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/2-cyano-l-beta-homophenylalanine-hydrochloride/6760?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart