CH6371381430 Chem Impex International, Inc.

3-Cyano-L-b-homophenylalanine hydrochloride

  • MDL number MFCD01861075
  • Cas Number : 270065-85-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 224-230
Synonym L-beta-HomoPhe(3-CN)-OH HCl
Purity 98
Storage temp. Store at 0-8
Physical Form White Powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/3-cyano-l-beta-homophenylalanine-hydrochloride/6763?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/3-cyano-l-beta-homophenylalanine-hydrochloride/6763?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart