CH6371381431 Chem Impex International, Inc.

4-Cyano-L-beta-homophenylalanine hydrochloride

  • MDL number MFCD01861078
  • Cas Number : 270065-88-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 212-218 °C
Purity 98
Storage temp. Store at 0-8°C
Physical Form White Powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/4-cyano-l-beta-homophenylalanine-hydrochloride/6766?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/4-cyano-l-beta-homophenylalanine-hydrochloride/6766?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart