CH6371381440 Chem Impex International, Inc.

Fmoc-(2-furyl)-D-b-homoalanine

  • MDL number MFCD01860994
  • Cas Number : 270596-34-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 139-145
Synonym Fmoc-(2-furyl)-D-beta-homoalanine
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-2-furyl-d-beta-homoalanine/6784?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-2-furyl-d-beta-homoalanine/6784?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart