CH6371381441 Chem Impex International, Inc.

Styryl-D-beta-homoalanine hydrochloride

  • MDL number MFCD01860995
  • Cas Number : 270596-35-5
  • Molecular Weight : 241.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 204-210 °C
Purity 98
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/styryl-d-beta-homoalanine-hydrochloride/6785?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/styryl-d-beta-homoalanine-hydrochloride/6785?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart