CH6371381443 Chem Impex International, Inc.

3-Chloro-L-b-homophenylalanine hydrochloride

  • MDL number MFCD01860998
  • Cas Number : 270596-38-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym L-beta-HomoPhe(3-Cl)-OH HCl
Purity 98
Storage temp. Store at 0-8
Physical Form White solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/3-chloro-l-ß-homophenylalanine-hydrochloride/6788?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/3-chloro-l-ß-homophenylalanine-hydrochloride/6788?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart