CH6371381444 Chem Impex International, Inc.

Boc-3-chloro-L-b-homophenylalanine

  • MDL number MFCD01860999
  • Cas Number : 270596-39-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 150-156 �C
Synonym Boc-L-beta-HomoPhe(3-Cl)-OH
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-3-chloro-l-beta-homophenylalanine/6789?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-3-chloro-l-beta-homophenylalanine/6789?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart