CH6371381448 Chem Impex International, Inc.

Fmoc-4-chloro-L-beta-homophenylalanine

  • MDL number MFCD01861002
  • Cas Number : 270596-43-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8°C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-4-chloro-l-beta-homophenylalanine/6793?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-4-chloro-l-beta-homophenylalanine/6793?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart