CH6371381455 Chem Impex International, Inc.

(S)-3-Amino-3-(3-bromophenyl)propionic acid

  • MDL number MFCD04113686
  • Cas Number : 275826-35-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-s-i-3-amino-3-3-bromophenyl-propionic-acid/6805?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-s-i-3-amino-3-3-bromophenyl-propionic-acid/6805?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart