CH6371381461 Chem Impex International, Inc.

2,4-Dichloro-D-b-homophenylalanine hydrochloride

  • MDL number MFCD01860945
  • Cas Number : 331847-13-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 203-209 �C
Synonym D-beta-HomoPhe(2,4-DiCl)-OH HCl
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/2-4-dichloro-d-beta-homophenylalanine-hydrochloride/6813?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/2-4-dichloro-d-beta-homophenylalanine-hydrochloride/6813?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart