CH6371381473 Chem Impex International, Inc.

(R)-a-(3-Chlorobenzyl)proline�

  • MDL number MFCD06797536
  • Cas Number : 637020-80-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 252-258
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-r-i-alpha-3-chlorobenzyl-proline-font-size-2-face-symbol-middot-font-hcl/6837?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-r-i-alpha-3-chlorobenzyl-proline-font-size-2-face-symbol-middot-font-hcl/6837?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart