CH6371381476 Chem Impex International, Inc.

Boc-(R)-a-benzylproline

  • MDL number MFCD06795522
  • Cas Number : 706806-60-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-i-r-i-a-benzylproline/6847?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-i-r-i-a-benzylproline/6847?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart