CH6371381479 Chem Impex International, Inc.

(+)-(1S,3R)-3-Amino-cyclopentane carboxylic acid

  • MDL number MFCD00211290
  • Cas Number : 71830-07-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (+)-(1S,3R)-beta-Homocycloleucine
Purity 98
Storage temp. Store at 0-8
Physical Form White solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/1-i-s-i-3-i-r-i-3-amino-cyclopentane-carboxylic-acid/6862?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/1-i-s-i-3-i-r-i-3-amino-cyclopentane-carboxylic-acid/6862?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart