CH6371381482 Chem Impex International, Inc.

(R)-3-Amino-3-(2-thienyl)propionic acid

  • MDL number MFCD04113649
  • Cas Number : 73495-10-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 220-225 �dec)
Synonym L-beta-Ala-(2-thienyl)-OH
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-r-i-3-amino-3-2-thienyl-propionic-acid/6865?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-r-i-3-amino-3-2-thienyl-propionic-acid/6865?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart