CH6371381484 Chem Impex International, Inc.

Fmoc-2,4-dichloro-L-phenylalanine

  • MDL number MFCD01863048
  • Cas Number : 352351-62-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 166-177 �C
Synonym Fmoc-L-Phe(2,4-DiCl)-OH
Purity 99
Storage temp. Store at 0-8
Physical Form White solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-2-4-dichloro-l-phenylalanine/6867?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-2-4-dichloro-l-phenylalanine/6867?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart