CH6371381486 Chem Impex International, Inc.

L-b-Homotyrosine hydrochloride

  • MDL number MFCD01862869
  • Cas Number : 336182-13-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym L-b-HomoTyr-OH�
Purity 0.95
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/l-b-homotyrosine-hydrochloride/6869?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/l-b-homotyrosine-hydrochloride/6869?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart