CH6371381490 Chem Impex International, Inc.

Fmoc-4-chloro-D-beta-homophenylalanine

  • MDL number MFCD01860904
  • Cas Number : 331763-60-1
  • Molecular Weight : 435.91

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8°C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-4-chloro-d-beta-homophenylalanine/6874?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-4-chloro-d-beta-homophenylalanine/6874?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart