CH6371381499 Chem Impex International, Inc.

Boc-2-bromo-D-b-homophenylalanine

  • MDL number MFCD03788797
  • Cas Number : 765263-36-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 110-116
Synonym Boc-D-beta-HomoPhe(2-Br)-OH
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-2-bromo-d-beta-homophenylalanine/6885?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-2-bromo-d-beta-homophenylalanine/6885?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart