CH6371381532 Chem Impex International, Inc.

(R)-3-Amino-3-(4-bromophenyl)propionic acid

  • MDL number MFCD03840386
  • Cas Number : 479074-63-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym D-beta-Phe(4-Br)-OH
Purity 0.98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-r-i-3-amino-3-4-bromophenyl-propionic-acid/6940?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-r-i-3-amino-3-4-bromophenyl-propionic-acid/6940?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart