CH6371381537 Chem Impex International, Inc.

Boc-(S)-3-amino-3-(3-trifluoromethylphenyl)propionic acid

  • MDL number MFCD03427926
  • Cas Number : 500770-78-5
  • Molecular Weight : 333.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8°C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-i-s-i-3-amino-3-3-trifluoromethylphenyl-propionic-acid/6950?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-i-s-i-3-amino-3-3-trifluoromethylphenyl-propionic-acid/6950?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart