CH6371381538 Chem Impex International, Inc.

(R)-3-Amino-3-(4-trifluoromethylphenyl)propionic acid

  • MDL number MFCD04113668
  • Cas Number : 774178-39-1
  • Molecular Weight : 233.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8°C
Physical Form White to off-white powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-r-i-3-amino-3-4-trifluoromethylphenyl-propionic-acid/6953?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-r-i-3-amino-3-4-trifluoromethylphenyl-propionic-acid/6953?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart