CH6371381540 Chem Impex International, Inc.

(S)-3-Amino-3-(3-cyanophenyl)propionic acid

  • MDL number MFCD04113688
  • Cas Number : 791778-00-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym L-beta-Phe(3-CN)-OH
Purity 98
Storage temp. Store at 0-8
Physical Form White solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-s-i-3-amino-3-3-cyanophenyl-propionic-acid/6960?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-s-i-3-amino-3-3-cyanophenyl-propionic-acid/6960?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart