CH6371381547 Chem Impex International, Inc.

Boc-(R)-3-amino-3-(2-nitrophenyl)propionic acid

  • MDL number MFCD03427969
  • Cas Number : 501015-23-2
  • Molecular Weight : 310.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store at 0-8°C
Physical Form White to off-white powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-i-r-i-3-amino-3-2-nitrophenyl-propionic-acid/6973?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-i-r-i-3-amino-3-2-nitrophenyl-propionic-acid/6973?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart